Tisk…

Anti-AKT1 Rabbit Polyclonal Antibody

Dodavatel: US Biological
A1125-11U
USBIA1125-11UEA 19740 CZK
USBIA1125-11U
Anti-AKT1 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR