Tisk…

Anti-Agouti-Related Protein Rabbit Polyclonal Antibody

Dodavatel: US Biological
A1059-63A
USBIA1059-63AEA 16520 CZK
USBIA1059-63A
Anti-Agouti-Related Protein Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Agouti-Related Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR