Tisk…

Anti-hADNP Rabbit Polyclonal Antibody

Dodavatel: US Biological
A0900-09
USBIA0900-09EA 14440 CZK
USBIA0900-09
Anti-hADNP Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: hADNP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR