Tisk…

Anti-TRIM59 Rabbit Polyclonal Antibody

Dodavatel: US Biological
138466
USBI138466EA 22330 CZK
USBI138466
Anti-TRIM59 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 5 ?g/ml; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: TRIM59 (tripartite motif containing 59)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR