Tisk…

Anti-Glutamic Pyruvic Transaminase Rabbit Polyclonal Antibody

Dodavatel: US Biological
137999
USBI137999EA 22360 CZK
USBI137999
Anti-Glutamic Pyruvic Transaminase Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 2.5 ?g/ml; Immunochemistry: 4-8?g/ml

Type: Primary
Antigen: Glutamic Pyruvic Transaminase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR