Tisk…

Anti-CALR Rabbit Polyclonal Antibody

Dodavatel: US Biological
136763
USBI136763EA 10110 CZK
USBI136763
Anti-CALR Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Western Blot: 1:5000-10000

Type: Primary
Antigen: CALR (calreticulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Drosophila, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR