Tisk…

Anti-Creatine Kinase Mouse Monoclonal Antibody [clone: 13F36]

Dodavatel: US Biological
136578
USBI136578EA 20310 CZK
USBI136578
Anti-Creatine Kinase Mouse Monoclonal Antibody [clone: 13F36]
Protilátky
Type: Primary
Antigen: Creatine Kinase
Clonality: monoclonal
Clone: 13F36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR