Tisk…

Anti-SPAG4 Mouse Monoclonal Antibody [clone: 3C8]

Dodavatel: US Biological
133740
USBI133740EA 18650 CZK
USBI133740
Anti-SPAG4 Mouse Monoclonal Antibody [clone: 3C8]
Protilátky
Type: Primary
Antigen: SPAG4 (sperm associated antigen 4)
Clonality: monoclonal
Clone: 3C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR