Tisk…

Anti-PLK3 Rabbit Polyclonal Antibody

Dodavatel: US Biological
131482
USBI131482EA 19820 CZK
USBI131482
Anti-PLK3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PLK3 (polo-like kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR