Tisk…

Anti-PDHA2 Mouse Polyclonal Antibody

Dodavatel: US Biological
131073
USBI131073EA 17740 CZK
USBI131073
Anti-PDHA2 Mouse Polyclonal Antibody
Protilátky
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: PDHA2 (pyruvate dehydrogenase (lipoamide) alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR