Tisk…

Anti-GLAST Rabbit Polyclonal Antibody

Dodavatel: US Biological
127335
USBI127335EA 18850 CZK
USBI127335
Anti-GLAST Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GLAST
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR