Tisk…

Anti-GATM Mouse Polyclonal Antibody

Dodavatel: US Biological
127192
USBI127192EA 18650 CZK
USBI127192
Anti-GATM Mouse Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GATM (glycine amidinotransferase (L-arginine:glycine amidinotransferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR