Tisk…

Anti-DNASE2 Mouse Polyclonal Antibody

Dodavatel: US Biological
125960
USBI125960EA 18650 CZK
USBI125960
Anti-DNASE2 Mouse Polyclonal Antibody
Protilátky
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: DNASE2 (deoxyribonuclease II, lysosomal)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR