Tisk…

Anti-ATAD3A Rabbit Polyclonal Antibody

Dodavatel: US Biological
123660
USBI123660EA 19820 CZK
USBI123660
Anti-ATAD3A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ATAD3A (ATPase family, AAA domain containing 3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR