Tisk…

Anti-XK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: US Biological
043918-AP
USBI043918-APEA 25170 CZK
USBI043918-AP
Anti-XK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: XK (X-linked Kx blood group (McLeod syndrome))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu XK
Název antigenu X-linked Kx blood group (McLeod syndrome)
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení AP (Alkaline Phosphatase)
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
Western blot Yes
ELISA Yes
Epitop N-terminal
Počet aminokyselin 82 to 111
Formulář liquid
Čištění purified by Protein A chromatography
Skladovací pufr PBS, pH 7,2, contains ~0,09% sodium azide
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR