Tisk…

Anti-UBA1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: US Biological
043501-APC
USBI043501-APCEA 25170 CZK
USBI043501-APC
Anti-UBA1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: UBA1 (ubiquitin-like modifier activating enzyme 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR