Tisk…

Anti-TRIM56 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: US Biological
043252-AP
USBI043252-APEA 25170 CZK
USBI043252-AP
Anti-TRIM56 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TRIM56 (tripartite motif containing 56)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR