Tisk…

Anti-TRIM41 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: US Biological
043239-FITC
USBI043239-FITCEA 25170 CZK
USBI043239-FITC
Anti-TRIM41 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TRIM41 (tripartite motif containing 41)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR