Tisk…

Anti-Staphylococcus aureus, delta Hemolysin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: US Biological
042370-FITC
USBI042370-FITCEA 25170 CZK
USBI042370-FITC
Anti-Staphylococcus aureus, delta Hemolysin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended dilution: Western Blot: 1:1000
Do not freeze!

Type: Primary
Antigen: Staphylococcus aureus, delta Hemolysin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR