Tisk…

Anti-SIX2 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
041765-BIOTIN
USBI041765-BIOTINEA 25170 CZK
USBI041765-BIOTIN
Anti-SIX2 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SIX2 (SIX homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR