Tisk…

Anti-PDHA1 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
039828-BIOTIN
USBI039828-BIOTINEA 25170 CZK
USBI039828-BIOTIN
Anti-PDHA1 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PDHA1 (pyruvate dehydrogenase (lipoamide) alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR