Tisk…

Anti-PABPN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: US Biological
039602-FITC
USBI039602-FITCEA 25170 CZK
USBI039602-FITC
Anti-PABPN1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: PABPN1 (poly(A) binding protein, nuclear 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR