Tisk…

Anti-NOP16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: US Biological
039055-HRP
USBI039055-HRPEA 25170 CZK
USBI039055-HRP
Anti-NOP16 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NOP16 (NOP16 nucleolar protein)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR