Tisk…

Anti-MAP3K1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: US Biological
038136-FITC
USBI038136-FITCEA 25170 CZK
USBI038136-FITC
Anti-MAP3K1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MAP3K1 (mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR