Tisk…

Anti-KCND1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: US Biological
037305-AP
USBI037305-APEA 25170 CZK
USBI037305-AP
Anti-KCND1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: KCND1 (potassium voltage-gated channel, Shal-related subfamily, member 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR