Tisk…

Anti-JAK2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: US Biological
037178-APC
USBI037178-APCEA 25170 CZK
USBI037178-APC
Anti-JAK2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR