Tisk…

Anti-ISL2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: US Biological
037122-APC
USBI037122-APCEA 25170 CZK
USBI037122-APC
Anti-ISL2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ISL2 (ISL LIM homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu ISL2
Environmentally Preferable
Název antigenu ISL LIM homeobox 2
Druh protilátky Primární
Klonovalita Polyclonal
Spojení APC (Allophycocyanin)
Reaktivita Human
Hostitel Rabbit
Izotyp IgG
Western blot Yes
ELISA Yes
Epitop N-terminal
Počet aminokyselin 1 to 30
Formulář liquid
Čištění purified by Protein A chromatography
Skladovací pufr PBS, pH 7,2, contains ~0,09% sodium azide
Skladovací teplota Cool
Transportní teplota Ice
Testované aplikace Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR