Tisk…

Anti-IL34 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: US Biological
037012-APC
USBI037012-APCEA 25170 CZK
USBI037012-APC
Anti-IL34 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: IL34 (interleukin 34)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR