Tisk…

Anti-HTATIP2 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
036818-BIOTIN
USBI036818-BIOTINEA 25170 CZK
USBI036818-BIOTIN
Anti-HTATIP2 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: HTATIP2 (HIV-1 Tat interactive protein 2, 30kDa)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR