Tisk…

Anti-ERBB2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: US Biological
035180-HRP
USBI035180-HRPEA 25170 CZK
USBI035180-HRP
Anti-ERBB2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR