Tisk…

Anti-DHRS2 Rabbit Polyclonal Antibody (Biotin)

Dodavatel: US Biological
034618-BIOTIN
USBI034618-BIOTINEA 25170 CZK
USBI034618-BIOTIN
Anti-DHRS2 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: DHRS2 (dehydrogenase/reductase (SDR family) member 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR