Tisk…

Anti-CELF6 Rabbit Polyclonal Antibody

Dodavatel: US Biological
032593
USBI032593EA 19000 CZK
USBI032593
Anti-CELF6 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: CELF6 (CUGBP, Elav-like family member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR