Tisk…

Anti-ACAP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: US Biological
031520-HRP
USBI031520-HRPEA 25170 CZK
USBI031520-HRP
Anti-ACAP1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: ACAP1 (ArfGAP with coiled-coil, ankyrin repeat and PH domains 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR