Tisk…

Anti-PRDM2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 33AT1045]

Dodavatel: US Biological
031203-FITC
USBI031203-FITCEA 25170 CZK
USBI031203-FITC
Anti-PRDM2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 33AT1045]
Protilátky
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PRDM2 (PR domain containing 2, with ZNF domain)
Clonality: Monoclonal
Clone: 33AT1045
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR