Tisk…

Anti-ROR1 Mouse Monoclonal Antibody [clone: 13B681]

Dodavatel: US Biological
030603
USBI030603EA 17830 CZK
USBI030603
Anti-ROR1 Mouse Monoclonal Antibody [clone: 13B681]
Protilátky
Type: Primary
Antigen: ROR1 (receptor tyrosine kinase-like orphan receptor 1)
Clonality: Monoclonal
Clone: 13B681
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR