Tisk…

Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody

Dodavatel: US Biological
0014-24E
USBI0014-24EEA 20160 CZK
USBI0014-24E
Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR