Tisk…

Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dodavatel: US Biological
0014-24C-PE
USBI0014-24C-PEEA 25170 CZK
USBI0014-24C-PE
Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Produktová specifikace

Symbol antigenu 14-3-3 delta/zeta
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Spojení PE (Phycoerythrin)
Reaktivita Human,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
ELISA Yes
Epitop C-terminal
Formulář liquid
Čištění purified by Protein G chromatography
Skladovací pufr PBS, pH 7,2
Skladovací teplota Cool
Transportní teplota Ice
Testované aplikace Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR