Tisk…
HPLC tubing, Tefzel® (ETFE)
Dodavatel: UPCHURCH SCIENTIFIC
1516 1516L 1516XL 1517 1517L 1517XL 1528L 1528XL 1529 1529L 1529XL 1530 1530L 1530XL 1528 1648 1648L 1648XL
UPCH1516EA 1690 CZK
UPCH1516 UPCH1516L 554-2959 UPCH1517 UPCH1517L UPCH1517XL UPCH1528L 554-2980 UPCH1529 UPCH1529L UPCH1529XL 554-2981 554-2982 554-2983 UPCH1528 UPCH1648 UPCH1648L 554-2991
HPLC tubing, Tefzel® (ETFE)
Hadičky Chromatography Tubing
Tefzel® (ETFE), natural.
  • Excellent solvent resistance
  • More durable and less gas permeable than PTFE, FEP and PFA
  • Operating temperatures up to 80 °C
ETFE tubing is chemically inert and suitable for higher pressure applications (when using aqueous mobile phase) than PTFE, FEP and PFA. Additionally, because Tefzel® is more rigid than PTFE, FEP and PFA, this tubing better resists inner diameter collapse.
¹/₁₆" ext. Ø tubing holds from 500 psi (34 bar) to 4000 psi (276 bar), depending upon the inner diameter; inner and outer diameter tolerances are ±0,00More Product Information