Tisk…
Electronic comparators, EComparator series
Dodavatel: Lovibond Tintometer
162010 163010 162020 163020
TINT162010EA 79770 CZK
TINT162010 TINT163010 TINT162020 TINT163020
Electronic comparators, EComparator series
Kolorimetry Sady porovnávací
The Lovibond® EComparator series provides an easy way to transition from subjective visual measurement to a non subjective, accurate electronic measurement.
  • Digital, portable, push-button technology
  • On-screen colour
  • On-screen numerical display
  • User friendly ergonomics and intuitive interface guarantee new users can be quickly trained and easily supported
  • Large data storage (>20000 readings)
  • USB connectivity ensures readings can be stored and shared easily and quickly
Informace o dodávce: Instrument supplied with USB cable, USB power supply, 4× AAMore Product Information