Tisk…
CryoMed™ Cryogenic Freezers with OPC UA, General Purpose
Dodavatel: Thermo Fisher Scientific
TSCM17SV TSCM34SV TSCM48SV TSCM17FV TSCM34FV
THBRTSCM17SVEA 1216380 CZK
THBRTSCM17SV THBRTSCM34SV THBRTSCM48SV THBRTSCM17FV THBRTSCM34FV
CryoMed™ Cryogenic Freezers with OPC UA, General Purpose
Mrazáky Mrazáky kryogenní
For customers in cell and gene therapy and vaccine production, the CryoMed with OPC UA provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed features OPC UA serial communication (ethernet) capabilities, comes standard factory certifications, provides enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
  • User-friendly operation with enhanced data traceability
  • Remote alarm contacts
  • Intuitive touchscreen display
More Product Information