Tisk…
Autoclave cleanroom labels, TX515A
Dodavatel: Texwipe
TX515A
TEXWTX515AEA 13830 CZK
TEXWTX515A
Autoclave cleanroom labels, TX515A
Štítky
These autoclavable labels are made of a Teslin® material designed for label printing and can be used with most conventional inks.
  • Matte finish
  • Permanent acrylic adhesive to provide excellent adhesion
  • Prevent the removal of written data during the autoclave process
Ideal for applications requiring label and label data to maintain integrity through autoclave process.