Tisk…
Gas detectors for carbon monoxides and dioxides, 315-3
Dodavatel: Testo
0632.3153
TEST0632.3153EA 26960 CZK
TEST0632.3153
Gas detectors for carbon monoxides and dioxides, 315-3
Systémy detekce úniku Systémy detekce úniku plynu
Escaping CO gases pose a risk of poisoning, the 315-3 constantly checks the level of CO and CO₂ in ambient air.
  • Data transfer to the Testo 330
  • Parallel mono and dioxyde measurement
  • Direct display of both gases
Certifikace: BS 6173:2009 and EN standard 50543.
Informace o dodávce: Supplied with USB mains unit and cable, batteries and instruction manual.