Tisk…
Multifunctional air velocity and indoor air quality (IAQ) instrument, testo 440 and Testo 440 differential pressure (dP)
Dodavatel: Testo
0563.4401 0563.4403 0560.4401 0560.4402
TEST0563.4401EA 15630 CZK
TEST0563.4401 TEST0563.4403 TEST0560.4401 620-2657
Multifunctional air velocity and indoor air quality (IAQ) instrument, testo 440 and Testo 440 differential pressure (dP)
Záznamníky dat (Dataloggery)
The testo 440 and testo 440 differential pressure (dP) combine the benefits of a compact handheld device with intuitive measurement menus and a comprehensive selection of air velocity and IAQ probes. This means that all measuring tasks on air conditioning and ventilation systems are reliably under control.
  • Just one instrument for all important air velocity and IAQ measurement demands
  • Clearly structured measurement menus for the most important applications stored on the instrument
  • Quick entering of application parameters
  • Wireless: Bluetooth probes for
More Product Information