Tisk…
Differential pressure manometers, testo 512
Dodavatel: Testo
0563.1512 0563.2512
TEST0563.1512EA 12760 CZK
TEST0563.1512 TEST0563.2512
Differential pressure manometers, testo 512
Manometry
This compact differential pressure measuring instrument testo 512 is the ideal solution for simple, fast and precise measurement of gas flow and static gas pressure on heating systems, for checking filters on air conditioning systems and for Pitot tube measurement in ventilation ducts.
  • Reliable measurement results due to position-independent differential pressure sensor with high accuracy
  • Fast in-app (duct) configuration, graph history, second screen and measurement data memory in the testo Smart App
  • Volume flow calculation in measuring instrument and App
More Product Information