Tisk…
SURPLUG™ AD Solution Administration Set
Dodavatel: TERUMO
SAQTF303NN
TERUSAQTF303NNEA 117380 CZK
TERUSAQTF303NN
SURPLUG™ AD Solution Administration Set
Infuze
SURPLUG™ AD series infusion system meeting the needs in various departments in a safer and simpler way.
  • A flat top for easier scrubbing
  • Structure for high flow rate
  • Applicable to blood transfusions
  • Round lock connector, PVC-free
  • AFF clip, SURPLUG AD T-Port×3
Using high-quality materials for the system, no need for a dedicated connection adaptor and Terumo's unique structure eliminates drug residues.