Tisk…
Four drawer storage cupboards
Dodavatel: TEKNOMEK
SHCBED10X60X20 SHCBED12X60X20 SHCBID10X60X20 SHCBID12X60X20
TEKMSHCBED10X60X20EA 0 CZK
TEKMSHCBED10X60X20 TEKMSHCBED12X60X20 TEKMSHCBID10X60X20 TEKMSHCBID12X60X20
Four drawer storage cupboards
Nábytek kancelářský Skříně pro zásoby
Our storage cupboard comes complete with four drawers to provide perfect storage.
  • Brushed dull polish finish
  • Sloping top
  • Four shelf height adjustments
  • Stainless steel adjustable feet with non-marking rubber base
  • Lockable main doors
  • Four internal or external drawers
  • Shelves sold separately
Our cupboard has been designed with a sloping top, four drawers (internal and external options available) and lockable doors to secure all content.
The range includes a single (600 mm) or double (1000 or 1200 mm) solid door.
Supplied with four easy clean shelfMore Product Information