Tisk…

Adenosine ≥99.0% (by HPLC, titration analysis)

Dodavatel: TCI

Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

A0152-25G A0152-5G
TCIAA0152-25GEA 2200 CZK
TCIAA0152-25G TCIAA0152-5G
Adenosine ≥99.0% (by HPLC, titration analysis)
Adenosine
Vzorec: C₁₀H₁₃N₅O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00005752
CAS číslo: 58-61-7
EINECS: 200-389-9
Merck Index: 14,00153
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8f98b56cc591e6ddd4e52847b503721638b6a81&vid=a8f98b56cc591e6ddd4e52847b503721638b6a81&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D -69.0 to -72.0 deg (C=2, NaOH sol. (5%))
Lambda max. 256.0 to 261.0 nm (0.1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR