Tisk…

Adenine sulphate dihydrate ≥98.0% (by HPLC, titration analysis)

Dodavatel: TCI
Varování

Synonyms: 6-Aminopurine sulphate dihydrate, Adenine hemisulphate dihydrate, 7H-Purin-6-amine hemisulphate dihydrate, 9H-Purin-6-amine hemisulphate dihydrate

A0151-25G
TCIAA0151-25GEA 4610 CZK
TCIAA0151-25G
Adenine sulphate dihydrate ≥98.0% (by HPLC, titration analysis)
Adenine sulphate dihydrate
Vzorec: (C₅H₅N₅)₂·H₂SO₄·2H₂O
MDL Number: MFCD00213655
CAS číslo: 6509-19-9
EINECS: 206-286-5
Merck Index: 14,00152

Objednat

Výsledky specifikačního testu

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on dried substance)
Solubility in hot Water almost transparency
Drying loss 8.0 to 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR