Tisk…

Oil red O, Supelco®

Dodavatel: Merck

Synonyms: CI 26125

O9755-25G
SUPLO9755-25GEA 2310 CZK
SUPLO9755-25G
Oil red O, Supelco®
Oil red O
Vzorec: C₂₆H₂₄N₄O
Storage Temperature: Ambient
MDL Number: MFCD00003898
CAS číslo: 1320-06-5
EINECS: 215-295-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR