Tisk…
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Dodavatel: Merck
39361-250ML
SUPL39361-250MLEA 1760 CZK
SUPL39361-250ML
Silver Standard Solution, for AAS, TraceCERT®, Supelco®
Standardy Standardy kationické Standardy - stříbro
This silver standard solution prepared with high quality silver metal in 2% HNO3.
Certifikace: ISO/IEC 17025 and ISO Guide 34. Traceable to NIST, SRM.